CAS RN 3383-96-8
IUPAC Name:
O-4-[(4-{[dimethoxy(sulfanylidene)-λ⁵-phosphanyl]oxy}phenyl)sulfanyl]phenyl O,O-dimethyl phosphorothioate
Synonyms:
abate diphos (pesticide) nephis caswell no. 845 nephis 1G temephos swebat american cyanamid CL-52160 procida abathion
Molecular Weight (g/mol):
466.46
Molecular Formula:
C16H20O6P2S3
InChi Key:
WWJZWCUNLNYYAU-UHFFFAOYSA-N
SMILES:
COP(=S)(OC)OC1=CC=C(SC2=CC=C(OP(=S)(OC)OC)C=C2)C=C1
1
–
1
of
1
results
1
Abate, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material