accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 338454-14-1

IUPAC Name: (1H-indazol-5-yl)boronic acid
Synonyms: 1H-indazol-5-ylboronic acid
Molecular Weight (g/mol): 161.96
Molecular Formula: C7H7BN2O2
InChi Key: CLVPGJWAMIADSY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2NN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results