CAS RN 33884-41-2
IUPAC Name:
3-(ethoxymethylidene)pentane-2,4-dione
Synonyms:
3-(ethoxymethylidene)pentane-2,4-dione
Molecular Weight (g/mol):
156.18
Molecular Formula:
C8H12O3
InChi Key:
ITTXGKOHFZJUEX-UHFFFAOYSA-N
SMILES:
CCOC=C(C(C)=O)C(C)=O
1
–
1
of
1
results
1
Promotions Available
3-Ethoxymethylene-2,4-pentanedione, 97+%
3-Ethoxymethylene-2,4-pentanedione, 97+%
CAS: 33884-41-2 Molecular Formula: C8H12O3 Molecular Weight (g/mol): 156.18 MDL Number: MFCD07368652 InChI Key: ITTXGKOHFZJUEX-UHFFFAOYSA-N PubChem CID: 240418 IUPAC Name: 3-(ethoxymethylidene)pentane-2,4-dione SMILES: CCOC=C(C(C)=O)C(C)=O