CAS RN 339-59-3
IUPAC Name:
4-(trifluoromethyl)benzohydrazide
Synonyms:
4-(trifluoromethyl)benzohydrazide
Molecular Weight (g/mol):
204.15
Molecular Formula:
C8H7F3N2O
InChi Key:
GKBDXTNCBPZMFX-UHFFFAOYSA-N
SMILES:
NNC(=O)C1=CC=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4-(Trifluoromethyl)benzhydrazide, 98%
4-(Trifluoromethyl)benzhydrazide, 98%
CAS: 339-59-3 Molecular Formula: C8H7F3N2O Molecular Weight (g/mol): 204.15 MDL Number: MFCD00051703 InChI Key: GKBDXTNCBPZMFX-UHFFFAOYSA-N Synonym: 4-trifluoromethyl benzhydrazide,4-trifluoromethyl benzohydrazide,4-trifluoromethyl benzoic acid hydrazide,4-trifluoromethylbenzhydrazide,4-trifluoromethylbenzoic acid hydrazide,4-trifluoromethyl benzene-1-carbohydrazide,1-trifluoromethyl benzene-4-carbohydrazide,alpha,alpha,alpha-trifluoro-p-toluic acid hydrazide,benzoic acid, 4-trifluoromethyl-, hydrazide,pubchem4506 PubChem CID: 520662 IUPAC Name: 4-(trifluoromethyl)benzohydrazide SMILES: NNC(=O)C1=CC=C(C=C1)C(F)(F)F