accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(S)HNOOCH3OOCH3CH3CH3

CAS RN 33900-28-6

IUPAC Name: methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Synonyms: methyl (2S)-2-[(tert-butoxycarbonyl)amino]-3-(1H-indol-3-yl)propanoate|BOC-TRP-OME
Molecular Weight (g/mol): 318.37
Molecular Formula: C17H22N2O4
InChi Key: QXLOVPXUZAOKBL-AWEZNQCLSA-N
SMILES: COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)