accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3NCS

CAS RN 33904-04-0

IUPAC Name: 4-isothiocyanato-1,2-dimethoxybenzene
Synonyms: 4-isothiocyanato-1,2-dimethoxybenzene
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: LHPZZVZPOZPDDB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)N=C=S
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)