accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCl

CAS RN 3391-10-4

IUPAC Name: 1-(4-chlorophenyl)ethan-1-ol
Synonyms: 1-(4-chlorophenyl)ethanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: MVOSNPUNXINWAD-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results