accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 339268-21-2

IUPAC Name: tetrapotassium hexacyanorutheniumbis(ylium)
Synonyms: tetrapotassium hexacyanorutheniumbis(ylium)
Molecular Weight (g/mol): 413.57
Molecular Formula: C6K4N6Ru
InChi Key: FULSJSKUKUZHLZ-UHFFFAOYSA-N
SMILES: [K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results