accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSBr

CAS RN 3393-78-0

IUPAC Name: 1-bromo-4-[(4-bromophenyl)sulfanyl]benzene
Synonyms: bis(4-bromophenyl) sulfide
Molecular Weight (g/mol): 344.06
Molecular Formula: C12H8Br2S
InChi Key: CLPVCAXOQZIJGW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(SC2=CC=C(Br)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results