accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 3395-91-3

IUPAC Name: methyl 3-bromopropanoate
Synonyms: methyl 3-bromopropanoate
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: KQEVIFKPZOGBMZ-UHFFFAOYSA-N
SMILES: COC(=O)CCBr
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (7)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)