accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClNH2HCl

CAS RN 33965-47-8

IUPAC Name: methyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride
Synonyms: methyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride
Molecular Weight (g/mol): 250.12
Molecular Formula: C10H13Cl2NO2
InChi Key: GCBCWTWQAFLKJG-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)