accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3397-29-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-(1,3-dioxoisoindol-2-yl)acetate
Molecular Weight (g/mol): 302.24
Molecular Formula: C14H10N2O6
InChi Key: NQZZTHXHURDXOR-UHFFFAOYSA-N
SMILES: O=C(CN1C(=O)C2=CC=CC=C2C1=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)