accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NNOHSOOO

CAS RN 33986-15-1

IUPAC Name: (cyanomethyl)(methyl)azanium hydrogen sulfate
Synonyms: (cyanomethyl)(methyl)azanium hydrogen sulfate
Molecular Weight (g/mol): 168.17
Molecular Formula: C3H8N2O4S
InChi Key: SEBZVAFMXLJJNO-UHFFFAOYSA-N
SMILES: C[NH2+]CC#N.OS([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results