accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3399-73-3

IUPAC Name: 2-(cyclohex-1-en-1-yl)ethan-1-amine
Synonyms: 2-(cyclohex-1-en-1-yl)ethanamine
Molecular Weight (g/mol): 125.22
Molecular Formula: C8H15N
InChi Key: IUDMXOOVKMKODN-UHFFFAOYSA-N
SMILES: NCCC1=CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results