accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 339985-36-3

IUPAC Name: 4-(5-bromothiophen-2-yl)-N,N-bis[4-(5-bromothiophen-2-yl)phenyl]aniline
Synonyms: 4-(5-bromothiophen-2-yl)-N,N-bis[4-(5-bromothiophen-2-yl)phenyl]aniline
Molecular Weight (g/mol): 728.37
Molecular Formula: C30H18Br3NS3
InChi Key: QWJSFRTXYYGQPZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=C(Br)S1)C1=CC=C(C=C1)C1=CC=C(Br)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results