accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3401-74-9

IUPAC Name: didodecyldimethylazanium chloride
Synonyms: dilauryldimonium chloride
Molecular Weight (g/mol): 418.19
Molecular Formula: C26H56ClN
InChi Key: WLCFKPHMRNPAFZ-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results