accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNOOOHNS(E/Z)

CAS RN 340316-62-3

IUPAC Name: 1-(3-bromophenyl)-5-[(furan-2-yl)methylidene]-2-sulfanylidene-1,3-diazinane-4,6-dione
Synonyms: 1-(3-bromophenyl)-5-(furan-2-ylmethylidene)-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular Weight (g/mol): 377.21
Molecular Formula: C15H9BrN2O3S
InChi Key: MVFJHEQDISFYIS-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N1C(=S)NC(=O)C(=CC2=CC=CO2)C1=O
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results