accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3C(E)

CAS RN 3404-55-5

IUPAC Name: (2E)-4-methylhex-2-ene
Synonyms: 2-hexene, 4-methyl-, (E)-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: MBNDKEPQUVZHCM-GQCTYLIANA-N
SMILES: CCC(C)\C=C\C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results