accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3404-61-3

H3CCH3CH2

CAS RN 3404-61-3

IUPAC Name: 3-methylhex-1-ene
Synonyms: 3-methyl-1-hexene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: RITONZMLZWYPHW-UHFFFAOYNA-N
SMILES: CCCC(C)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results