accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34040-62-5

IUPAC Name: methyl 2-(chloromethyl)benzoate
Synonyms: methyl 2-(chloromethyl)benzoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: AAHZCIWUDPKSJP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results