accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3O

CAS RN 34047-39-7

IUPAC Name: 4-(methylsulfanyl)butan-2-one
Synonyms: 4-(methylsulfanyl)butan-2-one
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: DRGHCRKOWMAZAO-UHFFFAOYSA-N
SMILES: CSCCC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results