accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNO

CAS RN 34052-90-9

IUPAC Name: 2-[3-(4,5-dihydro-1,3-oxazol-2-yl)phenyl]-4,5-dihydro-1,3-oxazole
Synonyms: 2-[3-(4,5-dihydro-1,3-oxazol-2-yl)phenyl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: HMOZDINWBHMBSQ-UHFFFAOYSA-N
SMILES: C1CN=C(O1)C1=CC(=CC=C1)C1=NCCO1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)