accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34054-55-2

IUPAC Name: yttrium(3+) phosphate
Synonyms: yttrium phosphate|yttrium(III) phosphate
Molecular Weight (g/mol): 183.88
Molecular Formula: O4PY
InChi Key: UXBZSSBXGPYSIL-UHFFFAOYSA-K
SMILES: [Y+3].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results