accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOOSClOO

CAS RN 3406-84-6

IUPAC Name: [1,1'-biphenyl]-4,4'-disulfonyl dichloride
Synonyms: [1,1'-biphenyl]-4,4'-disulfonyl dichloride
Molecular Weight (g/mol): 351.21
Molecular Formula: C12H8Cl2O4S2
InChi Key: OTFAWEIFBPUXOH-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)