CAS RN 34084-50-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ 7-Aminoflunitrazepam Solution, 1.0 mg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
This certified solution standard is suitable for numerous GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.