accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 341-23-1

IUPAC Name: 4-fluoro-1H-indazole
Synonyms: 4-fluoro-1H-indazole
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: YTNDJUBXSVSRQK-UHFFFAOYSA-N
SMILES: FC1=C2C=NNC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results