CAS RN 34102-49-3
IUPAC Name:
(1R,6S)-7-azabicyclo[4.2.0]octan-8-one
Synonyms:
(1R,6S)-7-azabicyclo[4.2.0]octan-8-one
Molecular Weight (g/mol):
125.17
Molecular Formula:
C7H11NO
InChi Key:
SQHJLZVUXVHXKC-RITPCOANSA-N
SMILES:
O=C1N[C@H]2CCCC[C@@H]12
1
–
1
of
1
results
1
7-Azabicyclo[4.2.0]octan-8-one, 98%, Thermo Scientific™
CAS: 34102-49-3 Molecular Formula: C7H11NO Molecular Weight (g/mol): 125.171 MDL Number: MFCD01863278 InChI Key: SQHJLZVUXVHXKC-UHFFFAOYSA-N Synonym: 7-azabicyclo 4.2.0 octan-8-one,7-azabicyclo 4,2,0 octan-8-one,7-aza-bicyclo 4.2.0 octan-8-one,7-azabicyclo 4.2.0 octane-8-one,7-azabicyclo 4.2.0 octan-8-one # PubChem CID: 549080 IUPAC Name: 7-azabicyclo[4.2.0]octan-8-one SMILES: C1CCC2C(C1)C(=O)N2