accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNS(E)

CAS RN 3411-95-8

IUPAC Name: 6-[(2E)-2,3-dihydro-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: 6-[(2E)-3H-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 227.28
Molecular Formula: C13H9NOS
InChi Key: NSDGEQWRYXOZLN-UKTHLTGXSA-N
SMILES: O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)