accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3HNONCH3H3C

CAS RN 34123-59-6

IUPAC Name: 3,3-dimethyl-1-[4-(propan-2-yl)phenyl]urea
Synonyms: tolkan isoproturon arelon ipu stefes isoproturon-50 belgran graminon hytane 500L arelon R IP-flo
Molecular Weight (g/mol): 206.29
Molecular Formula: C12H18N2O
InChi Key: PUIYMUZLKQOUOZ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(NC(=O)N(C)C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results