accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34156-56-4

IUPAC Name: trisodium hexahydrate phosphonatoformate
Synonyms: foscarnet sodium hexahydrate trisodium phosphonoformate hexahydrate
Molecular Weight (g/mol): 300.04
Molecular Formula: CH12Na3O11P
InChi Key: ILRVASBWNRYBFD-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results