accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSOOOK

CAS RN 34169-62-5

IUPAC Name: potassium 1,4-dioxo-1,4-dihydronaphthalene-2-sulfonate
Synonyms: potassium 1,4-dioxonaphthalene-2-sulfonate
Molecular Weight (g/mol): 276.3
Molecular Formula: C10H5KO5S
InChi Key: WZEGLRXCWYMIDO-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)C1=CC(=O)C2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results