accessibility menu, dialog, popup

UserName

Viewing profile as user:
FO

CAS RN 342-24-5

IUPAC Name: (2-fluorophenyl)(phenyl)methanone
Synonyms: (2-fluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 200.21
Molecular Formula: C13H9FO
InChi Key: DWFDQVMFSLLMPE-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)