accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3420-72-2

IUPAC Name: (2E)-1-(2-hydroxy-4,6-dimethoxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
Synonyms: flavokavain A flavokawin A
Molecular Weight (g/mol): 314.34
Molecular Formula: C18H18O5
InChi Key: CGIBCVBDFUTMPT-RMKNXTFCSA-N
SMILES: COC1=CC=C(\C=C\C(=O)C2=C(O)C=C(OC)C=C2OC)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results