accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClCl(E)

CAS RN 34209-97-7

IUPAC Name: 2,4-dichloro-1-[(1E)-2-nitroethenyl]benzene
Synonyms: 2,4-dichloro-B-nitrostyrene
Molecular Weight (g/mol): 218.03
Molecular Formula: C8H5Cl2NO2
InChi Key: LIWIJBBAMBDXME-ONEGZZNKSA-N
SMILES: [O-][N+](=O)\C=C\C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results