accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOONHNHOOOHO

CAS RN 34235-82-0

IUPAC Name: 6-{[(benzyloxy)carbonyl]amino}-2-(4-methylbenzenesulfonamido)hexanoic acid
Synonyms: 6-{[(benzyloxy)carbonyl]amino}-2-(4-methylbenzenesulfonamido)hexanoic acid
Molecular Weight (g/mol): 434.51
Molecular Formula: C21H26N2O6S
InChi Key: XSWGRGGVKUEBSE-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)