CAS RN 342404-46-0
CAS RN 342404-46-0
IUPAC Name:
[1-(benzenesulfonyl)-1H-indol-2-yl]boronic acid
Synonyms:
1-(benzenesulfonyl)indol-2-ylboronic acid
Molecular Weight (g/mol):
301.12
Molecular Formula:
C14H12BNO4S
InChi Key:
HXWLCYMHOULBJZ-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid, 97%, Thermo Scientific™
CAS: 342404-46-0 Molecular Formula: C14H12BNO4S Molecular Weight (g/mol): 301.12 MDL Number: MFCD03086094 InChI Key: HXWLCYMHOULBJZ-UHFFFAOYSA-N Synonym: 1-phenylsulfonyl-2-indoleboronic acid,1-phenylsulfonyl-1h-indol-2-yl boronic acid,1-phenylsulfonyl-1h-indol-2-ylboronic acid,1-phenylsulfonyl-2-indolylboronic acid,1-phenylsulfonyl indole-2-boronic acid,1-benzenesulfonyl indol-2-ylboronic acid,boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl,1-benzenesulfonyl indol-2-yl boronic acid,acmc-209i5v,1-phenylsulfonylindole-2-boronic acid PubChem CID: 2776228 IUPAC Name: [1-(benzenesulfonyl)indol-2-yl]boronic acid SMILES: OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1