accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34246-96-3

IUPAC Name: 1-methoxypyrene
Synonyms: 1-methoxypyrene
Molecular Weight (g/mol): 232.28
Molecular Formula: C17H12O
InChi Key: CPPFVJLHDNOVTN-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results