accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34252-54-5

IUPAC Name: 2-methyl-2H-indazole-3-carbaldehyde
Synonyms: 2-methylindazole-3-carbaldehyde
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: SMWLSZQHLMZHHV-UHFFFAOYSA-N
SMILES: CN1N=C2C=CC=CC2=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results