accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 342613-63-2

IUPAC Name: methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Synonyms: methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Molecular Weight (g/mol): 255.07
Molecular Formula: C9H7BrN2O2
InChi Key: VFFFBWRUZMAHFS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=NC=C(Br)N2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results