accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 343262-51-1

IUPAC Name: 2-bromo-5-methanesulfonylpyridine
Synonyms: 2-bromo-5-methanesulfonylpyridine
Molecular Weight (g/mol): 236.08
Molecular Formula: C6H6BrNO2S
InChi Key: ZRXQHWYUAIXKRL-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CN=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results