accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3433-80-5

IUPAC Name: 1-bromo-2-(bromomethyl)benzene
Synonyms: 1-bromo-2-(bromomethyl)benzene
Molecular Weight (g/mol): 249.93
Molecular Formula: C7H6Br2
InChi Key: LZSYGJNFCREHMD-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)