accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 343338-28-3

IUPAC Name: 2-methylpropane-2-sulfinamide
Synonyms: tert-butanesulfinamide
Molecular Weight (g/mol): 121.2
Molecular Formula: C4H11NOS
InChi Key: CESUXLKAADQNTB-UHFFFAOYNA-N
SMILES: CC(C)(C)S(N)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (3)
Quantity 
  • (4)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (4)