accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 343351-67-7

IUPAC Name: 2-(4-methylbenzenesulfonyl)-5-nitrofuran
Synonyms: 2-(4-methylbenzenesulfonyl)-5-nitrofuran
Molecular Weight (g/mol): 267.26
Molecular Formula: C11H9NO5S
InChi Key: GAUHIPWCDXOLCZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results