accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHHClHCl

CAS RN 34352-59-5

IUPAC Name: 1-methylpiperazine dihydrochloride
Synonyms: piperazine, 1-methyl- dihydrochloride
Molecular Weight (g/mol): 173.08
Molecular Formula: C5H14Cl2N2
InChi Key: AILFRWRYZZVJTL-UHFFFAOYSA-N
SMILES: Cl.Cl.CN1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results