accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClICl

CAS RN 343781-41-9

IUPAC Name: 3,5-dichloro-4-iodopyridine
Synonyms: 3,5-dichloro-4-iodopyridine
Molecular Weight (g/mol): 273.88
Molecular Formula: C5H2Cl2IN
InChi Key: DEXLTMBFLWCGNF-UHFFFAOYSA-N
SMILES: ClC1=CN=CC(Cl)=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results