accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHO

CAS RN 34381-71-0

IUPAC Name: (1-methylpyrrolidin-2-yl)methanol
Synonyms: (1-methylpyrrolidin-2-yl)methanol
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: VCOJPHPOVDIRJK-UHFFFAOYNA-N
SMILES: CN1CCCC1CO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)