accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COFFF

CAS RN 344-00-3

IUPAC Name: ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
Synonyms: ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
Molecular Weight (g/mol): 198.14
Molecular Formula: C7H9F3O3
InChi Key: YLRGPBKEZVHOAW-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)C(=O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results