accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOBrFFF

CAS RN 344-38-7

IUPAC Name: 4-bromo-1-nitro-2-(trifluoromethyl)benzene
Synonyms: 4-bromo-1-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 270.01
Molecular Formula: C7H3BrF3NO2
InChi Key: ZHLYHEDQTJZYFI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)C=C1C(F)(F)F
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)