accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34404-28-9

CH3CH3CH3OOHNOHOOHO

CAS RN 34404-28-9

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}pentanedioic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]pentanedioic acid
Molecular Weight (g/mol): 247.25
Molecular Formula: C10H17NO6
InChi Key: AQTUACKQXJNHFQ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)