accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNNOOHHOHOClN

CAS RN 34408-14-5

IUPAC Name: 2-(6-amino-8-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-(6-amino-8-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 301.69
Molecular Formula: C10H12ClN5O4
InChi Key: MHDPPLULTMGBSI-UHFFFAOYNA-N
SMILES: NC1=C2N=C(Cl)N(C3OC(CO)C(O)C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results